Assay Details
Target Gene Details
Entrez Gene ID: | 10914 |
Gene Name: | poly(A) polymerase alpha |
Gene Aliases: |
PAP |
Location: |
Chr.14:96502376-96567116 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAPOLA | NM_001252006.1 | NP_001238935.1 | ||
| NM_001252007.1 | NP_001238936.1 | |||
| NM_001293627.1 | NP_001280556.1 | |||
| NM_001293628.1 | NP_001280557.1 | |||
| NM_001293632.1 | NP_001280561.1 | |||
| NM_032632.4 | NP_116021.2 | |||
| XM_005267281.1 | XP_005267338.1 | |||
| XM_005267282.2 | XP_005267339.1 | |||
| XM_017020935.1 | XP_016876424.1 | |||
| XM_017020936.1 | XP_016876425.1 | |||
| AK295020.1 | ||||
| AK302406.1 | ||||
| AK307659.1 | ||||
| BC000927.1 | AAH00927.1 | |||
| BC036014.1 | AAH36014.1 | |||
| BX161482.1 | CAD61935.1 | |||
| BX248301.1 | CAD62628.1 | |||
| BX248753.1 | CAD66560.1 | |||
| DB045100.1 | ||||
| DB483858.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1052499 | Chr.14:96508187 - 96568468 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map