Assay Details
Target Gene Details
Entrez Gene ID: | 122481 |
Gene Name: | adenylate kinase 7 |
Gene Aliases: |
AK 7 |
Location: |
Chr.14:96392111-96489427 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AK7 | NM_152327.3 | NP_689540.2 | ||
| XM_006720021.2 | XP_006720084.1 | |||
| XM_006720022.2 | XP_006720085.1 | |||
| XM_011536403.2 | XP_011534705.1 | |||
| XM_017020956.1 | XP_016876445.1 | |||
| XM_017020957.1 | XP_016876446.1 | |||
| XM_017020958.1 | XP_016876447.1 | |||
| XM_017020959.1 | XP_016876448.1 | |||
| XM_017020960.1 | XP_016876449.1 | |||
| AK057426.1 | BAB71480.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3635423 | Chr.14:96385068 - 96403803 on Build GRCh38 | Gain |
|
| nsv565649 | Chr.14:96377253 - 96453283 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map