Assay Details
Target Gene Details
Entrez Gene ID: | 10979 |
Gene Name: | fermitin family member 2 |
Gene Aliases: |
KIND2, MIG2, PLEKHC1, UNC112, UNC112B, mig-2 |
Location: |
Chr.14:52857271-52951192 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FERMT2 | NM_001134999.1 | NP_001128471.1 | ||
| NM_001135000.1 | NP_001128472.1 | |||
| NM_006832.2 | NP_006823.1 | |||
| XM_005267285.2 | XP_005267342.1 | |||
| XM_006720008.2 | XP_006720071.1 | |||
| XM_006720009.2 | XP_006720072.1 | |||
| XM_006720010.2 | XP_006720073.1 | |||
| AF443279.1 | AAN75823.1 | |||
| AK291738.1 | ||||
| BC017327.2 | AAH17327.1 | |||
| BX161467.1 | CAD61925.1 | |||
| CR749486.1 | 1 | 862 | ||
| EU979385.1 | ||||
| Z24725.1 | CAA80852.1 |
More Information
Set Membership: |
|

Genomic Map