Assay Details
Target Gene Details
Entrez Gene ID: | 2103 |
Gene Name: | estrogen related receptor beta |
Gene Aliases: |
DFNB35, ERR2, ERRb, ESRL2, NR3B2 |
Location: |
Chr.14:76371347-76501839 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ESRRB | NM_004452.3 | NP_004443.3 | ||
| DR007211.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv565206 | Chr.14:76369614 - 76547692 on Build GRCh38 | Gain |
|
| dgv1942n100 | Chr.14:76358893 - 76552223 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map