Assay Details
Target Gene Details
Entrez Gene ID: | 283635 |
Gene Name: | family with sequence similarity 177 member A1 |
Gene Aliases: |
C14orf24 |
Location: |
Chr.14:35044907-35083383 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM177A1 | NM_001079519.1 | NP_001072987.1 | ||
| NM_001289022.1 | NP_001275951.1 | |||
| NM_173607.3 | NP_775878.2 | |||
| AK096173.1 | BAC04716.1 | |||
| AL833508.1 | ||||
| BC029559.1 | AAH29559.1 | |||
| BG700755.1 | ||||
| BX640884.1 | ||||
| BX640984.1 | ||||
| CR749796.1 | CAH18656.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564263 | Chr.14:34997378 - 35087292 on Build GRCh38 | Loss |
|
| esv3892499 | Chr.14:34900273 - 35107577 on Build GRCh38 | Loss |
|
| nsv564264 | Chr.14:35013591 - 35263163 on Build GRCh38 | Gain |
|
| nsv564265 | Chr.14:35043129 - 35155625 on Build GRCh38 | Loss |
|
| dgv1868n100 | Chr.14:35041579 - 35287111 on Build GRCh38 | Gain |
|
| esv3584249 | Chr.14:35060670 - 35255038 on Build GRCh38 | Gain |
|
| nsv1042625 | Chr.14:35076888 - 35211152 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115825077] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map