Assay Details
Target Gene Details
Entrez Gene ID: | 254028 |
Gene Name: | uncharacterized LOC254028 |
Gene Aliases: |
- |
Location: |
Chr.14:20588224-20607221 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC254028 | NR_122043.1 | |||
| BI830942.1 | ||||
| DB034207.1 | ||||
| DB053670.1 |
Target Gene Details
Entrez Gene ID: | 122651 |
Gene Name: | ribonuclease A family member 11 (inactive) |
Gene Aliases: |
C14orf6, HEL-S-84p, RAJ1 |
Location: |
Chr.14:20582892-20590258 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNASE11 | NM_145250.3 | NP_660293.1 | ||
| AY358794.1 | AAQ89154.1 | |||
| BX161458.1 | CAD61920.1 | |||
| CB995247.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv23438 | Chr.14:20332606 - 20789719 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75797650] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map