Assay Details
Target Gene Details
Entrez Gene ID: | 9692 |
Gene Name: | KIAA0391 |
Gene Aliases: |
MRPP3, PRORP |
Location: |
Chr.14:35121855-35274078 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIAA0391 | NM_001256678.1 | NP_001243607.1 | ||
| NM_001256679.1 | NP_001243608.1 | |||
| NM_001256680.1 | NP_001243609.1 | |||
| NM_001256681.1 | NP_001243610.1 | |||
| NM_014672.3 | NP_055487.2 | |||
| XM_005268237.3 | XP_005268294.1 | |||
| XM_011537409.2 | XP_011535711.1 | |||
| XM_011537410.2 | XP_011535712.1 | |||
| XM_017021835.1 | XP_016877324.1 | |||
| XM_017021836.1 | XP_016877325.1 | |||
| XM_017021837.1 | XP_016877326.1 | |||
| AB002389.1 | ||||
| AK301931.1 | ||||
| AK303508.1 | ||||
| AK304066.1 | ||||
| BC032221.1 | AAH32221.1 | |||
| BC044580.1 | AAH44580.1 | |||
| BP373829.1 | ||||
| BQ063741.1 | ||||
| BX161394.1 | CAD61881.1 | |||
| BX161487.1 | CAD61937.1 | |||
| DC404874.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564264 | Chr.14:35013591 - 35263163 on Build GRCh38 | Gain |
|
| nsv564265 | Chr.14:35043129 - 35155625 on Build GRCh38 | Loss |
|
| dgv1868n100 | Chr.14:35041579 - 35287111 on Build GRCh38 | Gain |
|
| esv3584249 | Chr.14:35060670 - 35255038 on Build GRCh38 | Gain |
|
| nsv1042625 | Chr.14:35076888 - 35211152 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map