Assay Details
Target Gene Details
Entrez Gene ID: | 4287 |
Gene Name: | ataxin 3 |
Gene Aliases: |
AT3, ATX3, JOS, MJD, MJD1, SCA3 |
Location: |
Chr.14:92058552-92106621 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATXN3 | NM_001127696.1 | 10 | 6753 | NP_001121168.1 |
| NM_001127697.2 | 9 | 6645 | NP_001121169.2 | |
| NM_001164774.1 | 4 | 6115 | NP_001158246.1 | |
| NM_001164776.1 | 5 | 6160 | NP_001158248.1 | |
| NM_001164777.1 | 4 | 5995 | NP_001158249.1 | |
| NM_001164778.1 | 7 | 6313 | NP_001158250.1 | |
| NM_001164779.1 | 7 | 6435 | NP_001158251.1 | |
| NM_001164780.1 | 8 | 6502 | NP_001158252.1 | |
| NM_001164781.1 | 9 | 6588 | NP_001158253.1 | |
| NM_001164782.1 | 3 | 5950 | NP_001158254.1 | |
| NM_004993.5 | 11 | 6798 | NP_004984.2 | |
| NM_030660.4 | 10 | 6633 | NP_109376.1 | |
| NR_028453.1 | 10 | 6712 | ||
| NR_028454.1 | 9 | 6547 | ||
| NR_028455.1 | 11 | 6766 | ||
| NR_028456.1 | 10 | 6601 | ||
| NR_028457.1 | 12 | 6857 | ||
| NR_028458.1 | 10 | 6701 | ||
| NR_028459.1 | 12 | 6852 | ||
| NR_028460.1 | 6 | 6227 | ||
| NR_028461.1 | 10 | 6710 | ||
| NR_028462.1 | 10 | 6689 | ||
| NR_028463.1 | 7 | 6401 | ||
| NR_028464.1 | 10 | 6699 | ||
| NR_028465.1 | 10 | 6721 | ||
| NR_028466.1 | 6 | 6347 | ||
| NR_028467.1 | 10 | 6713 | ||
| NR_028468.1 | 9 | 6545 | ||
| NR_028469.1 | 9 | 6559 | ||
| NR_028470.1 | 4 | 6017 | ||
| NR_031765.1 | 5 | 6214 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv565519 | Chr.14:92033004 - 92089711 on Build GRCh38 | Loss |
|
| dgv1956n100 | Chr.14:92056253 - 92104511 on Build GRCh38 | Gain |
|
| nsv1045760 | Chr.14:92030812 - 92090267 on Build GRCh38 | Gain |
|
| nsv527458 | Chr.14:91974722 - 92059127 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map