Assay Details
Target Gene Details
Entrez Gene ID: | 4707 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit B1 |
Gene Aliases: |
CI-MNLL, CI-SGDH, MNLL |
Location: |
Chr.14:92116123-92121809 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFB1 | NM_004545.3 | NP_004536.2 | ||
| AF050638.1 | AAD42054.1 | |||
| AK316557.2 | ||||
| BC009691.1 | AAH09691.2 | |||
| BC104672.1 | AAI04673.1 | |||
| BC126232.1 | ||||
| BC126234.1 | ||||
| BM052981.1 | ||||
| BX476462.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819246 | Chr.14:92113545 - 92121331 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map