Assay Details
Target Gene Details
Entrez Gene ID: | 55671 |
Gene Name: | protein phosphatase 4 regulatory subunit 3A |
Gene Aliases: |
FLFL1, KIAA2010, MSTP033, PP4R3, PP4R3A, SMEK1, smk-1, smk1 |
Location: |
Chr.14:91457481-91510554 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP4R3A | NM_001284280.1 | NP_001271209.1 | ||
| NM_001284281.1 | NP_001271210.1 | |||
| XM_005267842.2 | XP_005267899.1 | |||
| AB095930.1 | ||||
| AK000714.1 | BAA91338.1 | |||
| AK024297.1 | BAB14877.1 | |||
| BC038932.1 | AAH38932.1 | |||
| BC072409.1 | AAH72409.1 | |||
| BX248247.1 | CAD62575.1 |
More Information
Set Membership: |
|

Genomic Map