Assay Details
Target Gene Details
Entrez Gene ID: | 801 |
Gene Name: | calmodulin 1 |
Gene Aliases: |
CALML2, CAMI, CPVT4, DD132, LQT14, PHKD, caM |
Location: |
Chr.14:90396983-90408275 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CALM1 | NM_006888.4 | NP_008819.1 | ||
| AK289937.1 | ||||
| AU122722.1 | ||||
| BC000454.2 | AAH00454.1 | |||
| BC008597.1 | AAH08597.1 | |||
| BC011834.2 | AAH11834.1 | |||
| M27319.1 | AAA35635.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1069812 | Chr.14:90397655 - 90398256 on Build GRCh38 | Deletion |
|
| nsv1131072 | Chr.14:90397656 - 90398256 on Build GRCh38 | Deletion |
|
| esv3635309 | Chr.14:90382200 - 90607667 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map