Assay Details
Target Gene Details
Entrez Gene ID: | 10972 |
Gene Name: | transmembrane p24 trafficking protein 10 |
Gene Aliases: |
P24(DELTA), S31I125, S31III125, TMP21, Tmp-21-I, p23, p24d1 |
Location: |
Chr.14:75131468-75176646 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMED10 | NM_006827.5 | NP_006818.3 | ||
| AJ004913.1 | CAA06213.1 | |||
| AK222972.1 | BAD96692.1 | |||
| AK296800.1 | ||||
| AK310371.1 | ||||
| AK312384.1 | ||||
| AL832012.2 | CAD89913.1 | |||
| BC001496.2 | AAH01496.1 | |||
| BC001825.2 | AAH01825.1 | |||
| BX248754.1 | CAD66561.1 | |||
| BX504393.1 | ||||
| X97442.1 | CAA66071.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832833 | Chr.14:75100965 - 75304860 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs144192272] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map