Assay Details
Target Gene Details
Entrez Gene ID: | 624 |
Gene Name: | bradykinin receptor B2 |
Gene Aliases: |
B2R, BK-2, BK2, BKR2, BRB2 |
Location: |
Chr.14:96204798-96244329 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BDKRB2 | NM_000623.3 | NP_000614.1 | ||
| AK299316.1 | ||||
| AK311049.1 | ||||
| CR749338.1 | CAH18192.1 | |||
| DA096163.1 | ||||
| DC422332.1 | ||||
| M88714.1 | AAB02793.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3635417 | Chr.14:96186480 - 96223406 on Build GRCh38 | Gain |
|
| esv3635419 | Chr.14:96217831 - 96224236 on Build GRCh38 | Loss |
|
| nsv1416 | Chr.14:96217449 - 96252771 on Build GRCh38 | Insertion |
|
| nsv983853 | Chr.14:96221559 - 96224948 on Build GRCh38 | Duplication |
|
| nsv515629 | Chr.14:96217861 - 96222332 on Build GRCh38 | Loss |
|
| nsv1051786 | Chr.14:96213058 - 96244781 on Build GRCh38 | Gain |
|
| dgv262n27 | Chr.14:96219367 - 96222332 on Build GRCh38 | Loss |
|
| esv3635418 | Chr.14:96209831 - 96229962 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117207253] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map