Assay Details
Target Gene Details
Entrez Gene ID: | 64093 |
Gene Name: | SPARC related modular calcium binding 1 |
Gene Aliases: |
OAS |
Location: |
Chr.14:69879388-70032366 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMOC1 | NM_001034852.2 | NP_001030024.1 | ||
| NM_022137.5 | NP_071420.1 | |||
| XM_005267995.1 | XP_005268052.1 | |||
| XM_005267996.1 | XP_005268053.1 | |||
| AJ249900.1 | CAC10352.1 | |||
| AK289988.1 | ||||
| AK313063.1 | ||||
| BC008608.1 | AAH08608.1 | |||
| BC011548.1 | AAH11548.1 | |||
| DA805236.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv456336 | Chr.14:69615239 - 70107432 on Build GRCh38 | Gain |
|
| nsv832824 | Chr.14:69853922 - 70044684 on Build GRCh38 | Gain |
|
| esv3634847 | Chr.14:69893972 - 69900059 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map