Assay Details
Target Gene Details
Entrez Gene ID: | 4776 |
Gene Name: | nuclear factor of activated T-cells 4 |
Gene Aliases: |
NF-AT3, NF-ATC4, NFAT3 |
Location: |
Chr.14:24366911-24379604 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFATC4 | NM_001136022.2 | NP_001129494.1 | ||
| NM_001198967.2 | NP_001185896.1 | |||
| NM_001320043.1 | NP_001306972.1 | |||
| AK293873.1 | ||||
| AK309339.1 | ||||
| AK310445.1 | ||||
| DC403050.1 | ||||
| EU887632.1 | ||||
| EU887633.1 | ||||
| EU887634.1 | ||||
| EU887635.1 | ||||
| EU887636.1 | ||||
| EU887637.1 |
More Information
Set Membership: |
|

Genomic Map