Assay Details
Target Gene Details
Entrez Gene ID: | 57862 |
Gene Name: | zinc finger protein 410 |
Gene Aliases: |
APA-1, APA1 |
Location: |
Chr.14:73886615-73932288 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF410 | NM_001242924.1 | NP_001229853.1 | ||
| NM_001242926.1 | NP_001229855.1 | |||
| NM_001242927.1 | NP_001229856.1 | |||
| NM_001242928.1 | NP_001229857.1 | |||
| NM_021188.2 | NP_067011.1 | |||
| NR_040251.1 | ||||
| AK223261.1 | BAD96981.1 | |||
| AK293350.1 | ||||
| AK298307.1 | ||||
| AK298520.1 | ||||
| AK299138.1 | ||||
| AK301382.1 | ||||
| AK303834.1 | ||||
| AK304609.1 | ||||
| AK309957.1 | ||||
| AK315206.1 | ||||
| BC050683.1 | AAH50683.1 | |||
| U90919.1 | AAB51059.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1053281 | Chr.14:73518500 - 74050155 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map