Assay Details
Target Gene Details
Entrez Gene ID: | 4140 |
Gene Name: | microtubule affinity regulating kinase 3 |
Gene Aliases: |
CTAK1, KP78, PAR1A, Par-1a |
Location: |
Chr.14:103385364-103503831 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952258 | Chr.14:103386664 - 103405163 on Build GRCh38 | Duplication |
|
| nsv9168 | Chr.14:103065392 - 103446969 on Build GRCh38 | Loss |
|
| nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss |
|
| esv2676094 | Chr.14:103388564 - 103402701 on Build GRCh38 | Deletion |
|
| dgv422e214 | Chr.14:103355152 - 103437655 on Build GRCh38 | Gain |
|
| esv3635594 | Chr.14:103388531 - 103402677 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map