Assay Details
Target Gene Details
Entrez Gene ID: | 1734 |
Gene Name: | deiodinase, iodothyronine, type II |
Gene Aliases: |
5DII, D2, DIOII, SelY, TXDI2 |
Location: |
Chr.14:80197525-80231054 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIO2 | NM_000793.5 | NP_000784.2 | ||
| AK292210.1 |
Target Gene Details
Entrez Gene ID: | 100628307 |
Gene Name: | DIO2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.14:80211419-80455467 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIO2-AS1 | NR_038355.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045241 | Chr.14:79419018 - 80267858 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map