Assay Details
Target Gene Details
Entrez Gene ID: | 90141 |
Gene Name: | EF-hand calcium binding domain 11 |
Gene Aliases: |
C14orf143 |
Location: |
Chr.14:89794984-89954777 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFCAB11 | NM_001284267.1 | NP_001271196.1 | ||
| NM_001284269.1 | NP_001271198.1 | |||
| NM_145231.3 | NP_660274.1 | |||
| AK094740.1 | ||||
| AL832321.1 | CAD38616.1 | |||
| BC001787.1 | AAH01787.1 | |||
| BP371056.1 | ||||
| CF593330.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3635297 | Chr.14:89618440 - 89955557 on Build GRCh38 | Gain |
|
| esv3635302 | Chr.14:89814485 - 89817780 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117495048] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map