Assay Details
Target Gene Details
Entrez Gene ID: | 1113 |
Gene Name: | chromogranin A |
Gene Aliases: |
CGA |
Location: |
Chr.14:92922589-92935293 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHGA | NM_001275.3 | NP_001266.1 | ||
| NM_001301690.1 | NP_001288619.1 | |||
| XM_011536370.1 | XP_011534672.1 | |||
| AK223381.1 | BAD97101.1 | |||
| AK313757.1 | ||||
| BC001059.2 | AAH01059.1 | |||
| BC006459.1 | AAH06459.1 | |||
| BC009384.2 | AAH09384.2 | |||
| BT006869.1 | AAP35515.1 | |||
| BX248263.1 | CAD62591.1 | |||
| J03483.1 | AAA52017.1 | |||
| J03915.1 | AAA52018.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1400 | Chr.14:92928206 - 92973049 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map