Assay Details
Target Gene Details
Entrez Gene ID: | 114088 |
Gene Name: | tripartite motif containing 9 |
Gene Aliases: |
RNF91, SPRING |
Location: |
Chr.14:50975261-51105173 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIM9 | NM_015163.5 | NP_055978.4 | ||
| XM_005267301.3 | XP_005267358.1 | |||
| XM_005267302.3 | XP_005267359.1 | |||
| XM_006720018.3 | XP_006720081.1 | |||
| XM_011536389.2 | XP_011534691.1 | |||
| XM_017020943.1 | XP_016876432.1 | |||
| XM_017020944.1 | XP_016876433.1 | |||
| XM_017020945.1 | XP_016876434.1 | |||
| XM_017020946.1 | XP_016876435.1 | |||
| XM_017020947.1 | XP_016876436.1 | |||
| XM_017020948.1 | XP_016876437.1 | |||
| XM_017020949.1 | XP_016876438.1 | |||
| XM_017020950.1 | XP_016876439.1 | |||
| AK054781.1 | 1 | 541 | ||
| AK055388.1 | BAB70913.1 | |||
| BC063872.1 | AAH63872.1 | |||
| D87458.2 | BAA13398.2 | |||
| DA173440.1 |
More Information
Set Membership: |
|

Genomic Map