Assay Details
Target Gene Details
Entrez Gene ID: | 623 |
Gene Name: | bradykinin receptor B1 |
Gene Aliases: |
B1BKR, B1R, BDKRB2, BKB1R, BKR1, BRADYB1 |
Location: |
Chr.14:96256210-96264763 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BDKRB1 | NM_000710.3 | NP_000701.2 | ||
| AJ238044.1 | CAB45650.1 | |||
| AK291970.1 | ||||
| AK309528.1 | ||||
| CB130642.1 | ||||
| KJ950627.1 | ||||
| KJ950628.1 | ||||
| U30271.1 | AAD14824.1 | |||
| W49512.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3635420 | Chr.14:96224959 - 96319922 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map