Assay Details
Target Gene Details
Entrez Gene ID: | 390443 |
Gene Name: | ribonuclease A family member 9 (inactive) |
Gene Aliases: |
HEL128, RAK1, h461 |
Location: |
Chr.14:20556093-20560931 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNASE9 | NM_001001673.3 | NP_001001673.2 | ||
| NM_001110356.1 | NP_001103826.1 | |||
| NM_001110357.1 | NP_001103827.1 | |||
| NM_001110358.1 | NP_001103828.1 | |||
| NM_001110359.1 | NP_001103829.1 | |||
| NM_001110360.1 | NP_001103830.1 | |||
| NM_001110361.1 | NP_001103831.1 | |||
| NM_001289110.1 | NP_001276039.1 | |||
| AY907670.1 | ||||
| EF061296.1 | ||||
| EF061297.1 | ||||
| EF061298.1 | ||||
| EF061299.1 | ||||
| EF061300.1 | ||||
| EF061301.1 | ||||
| EF061302.1 | ||||
| EF061303.1 | 3 | 1159 | ||
| EF061304.1 | 4 | 1194 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv23438 | Chr.14:20332606 - 20789719 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73575070] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map