Assay Details
Target Gene Details
Entrez Gene ID: | 9321 |
Gene Name: | thyroid hormone receptor interactor 11 |
Gene Aliases: |
ACG1A, CEV14, GMAP-210, GMAP210, TRIP-11, TRIP230 |
Location: |
Chr.14:91965991-92040140 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIP11 | NM_001321851.1 | 21 | 7496 | NP_001308780.1 |
| NM_004239.4 | 21 | 7499 | NP_004230.2 | |
| XM_017021787.1 | 22 | 7520 | XP_016877276.1 | |
| XM_017021788.1 | 19 | 7371 | XP_016877277.1 | |
| BU628691.1 | 1 | 608 | ||
| BX486357.1 | 1 | 505 |
More Information
Set Membership: |
|

Genomic Map