Assay Details
Target Gene Details
Entrez Gene ID: | 97 |
Gene Name: | acylphosphatase 1 |
Gene Aliases: |
ACYPE |
Location: |
Chr.14:75053221-75069483 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACYP1 | NM_001107.4 | NP_001098.1 | ||
| NM_001302616.1 | NP_001289545.1 | |||
| NM_001302617.1 | NP_001289546.1 | |||
| NR_126393.1 | ||||
| NR_126394.1 | ||||
| NR_126395.1 | ||||
| AK312101.1 | ||||
| BC035568.1 | AAH35568.1 | |||
| BE467248.1 | ||||
| BG614847.1 | ||||
| BP316920.1 | ||||
| BX104912.1 | ||||
| CD675334.1 | ||||
| DB071115.1 | ||||
| HY012032.1 | ||||
| X84194.1 | CAA58987.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3634953 | Chr.14:75054238 - 75058816 on Build GRCh38 | Loss |
|
| esv3551775 | Chr.14:75054195 - 75058816 on Build GRCh38 | Deletion |
|
| nsv565188 | Chr.14:75053362 - 75055876 on Build GRCh38 | Loss |
|
| dgv3778n54 | Chr.14:75053172 - 75058746 on Build GRCh38 | Loss |
|
| dgv3779n54 | Chr.14:75054287 - 75055876 on Build GRCh38 | Loss |
|
| nsv510644 | Chr.14:75045880 - 75060238 on Build GRCh38 | Deletion |
|
| esv29535 | Chr.14:75054256 - 75058745 on Build GRCh38 | Loss |
|
| esv2677383 | Chr.14:75054235 - 75058816 on Build GRCh38 | Deletion |
|
| dgv1103n106 | Chr.14:75054197 - 75058897 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map