Assay Details
Target Gene Details
Entrez Gene ID: | 3429 |
Gene Name: | interferon alpha inducible protein 27 |
Gene Aliases: |
FAM14D, ISG12, ISG12A, P27 |
Location: |
Chr.14:94110733-94116699 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892577 | Chr.14:94096847 - 94118727 on Build GRCh38 | Gain |
|
| esv34067 | Chr.14:94024355 - 94160851 on Build GRCh38 | Loss |
|
| esv3635373 | Chr.14:94040151 - 94173989 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74536787] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map