Assay Details
Target Gene Details
Entrez Gene ID: | 5083 |
Gene Name: | paired box 9 |
Gene Aliases: |
STHAG3 |
Location: |
Chr.14:36657568-36677807 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAX9 | NM_006194.3 | NP_006185.1 | ||
| AB248958.1 | BAE79273.1 | |||
| AK310339.1 | ||||
| AK313627.1 | ||||
| BC001159.1 | AAH01159.1 | |||
| U59628.1 | AAD09487.1 | |||
| X92850.1 | CAA63436.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs77819352] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map