Assay Details
Target Gene Details
Entrez Gene ID: | 125228 |
Gene Name: | family with sequence similarity 210 member A |
Gene Aliases: |
C18orf19, HsT2329 |
Location: |
Chr.18:13663347-13726592 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM210A | NM_001098801.1 | 5 | 2814 | NP_001092271.1 |
| NM_152352.3 | 4 | 2674 | NP_689565.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv576529 | Chr.18:13606185 - 14150725 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115664532] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map