Assay Details
Target Gene Details
Entrez Gene ID: | 125228 |
Gene Name: | family with sequence similarity 210 member A |
Gene Aliases: |
C18orf19, HsT2329 |
Location: |
Chr.18:13663347-13726592 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM210A | NM_001098801.1 | NP_001092271.1 | ||
| NM_152352.3 | NP_689565.2 | |||
| AK055618.1 | BAB70973.1 | |||
| AK309506.1 | ||||
| AK309819.1 | ||||
| BU189118.1 | ||||
| DA573992.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv28578 | Chr.18:13672295 - 13676729 on Build GRCh38 | Loss |
|
| nsv821605 | Chr.18:13672884 - 13676729 on Build GRCh38 | Deletion |
|
| nsv576529 | Chr.18:13606185 - 14150725 on Build GRCh38 | Gain |
|

Genomic Map