Assay Details
Target Gene Details
Entrez Gene ID: | 8193 |
Gene Name: | double PHD fingers 1 |
Gene Aliases: |
BAF45b, NEUD4, neuro-d4 |
Location: |
Chr.19:38211006-38229714 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPF1 | NM_001135155.2 | NP_001128627.1 | ||
| NM_001135156.2 | NP_001128628.1 | |||
| NM_001289978.1 | NP_001276907.1 | |||
| NM_004647.3 | NP_004638.2 | |||
| XM_005259289.2 | XP_005259346.1 | |||
| XM_005259290.3 | XP_005259347.1 | |||
| XM_005259292.4 | XP_005259349.1 | |||
| XM_006723407.2 | XP_006723470.1 | |||
| XM_006723408.3 | XP_006723471.1 | |||
| XM_006723409.2 | XP_006723472.1 | |||
| XM_011527356.1 | XP_011525658.1 | |||
| XM_011527357.2 | XP_011525659.1 | |||
| XM_011527358.2 | XP_011525660.1 | |||
| AK297581.1 | ||||
| AK308563.1 | ||||
| AL529673.3 | ||||
| AL538568.3 | ||||
| BC021191.2 | AAH21191.1 | |||
| BC125152.1 | ||||
| BC125153.1 | ||||
| GQ358728.1 | ||||
| U43843.1 | AAC50685.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833821 | Chr.19:38139297 - 38266152 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map