Assay Details
Target Gene Details
Entrez Gene ID: | 4713 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit B7 |
Gene Aliases: |
B18, CI-B18 |
Location: |
Chr.19:14566078-14572077 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFB7 | NM_004146.5 | NP_004137.2 | ||
| AF112200.1 | AAF17188.1 | |||
| AF217091.1 | AAF91223.1 | |||
| BC002595.2 | AAH02595.1 | |||
| CB124371.1 | ||||
| CR450354.1 | CAG29350.1 | |||
| M33374.1 | AAA35675.1 |
More Information
Set Membership: |
|

Genomic Map