Assay Details
Target Gene Details
Entrez Gene ID: | 6141 |
Gene Name: | ribosomal protein L18 |
Gene Aliases: |
L18 |
Location: |
Chr.19:48615327-48619418 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL18 | NM_000979.3 | NP_000970.1 | ||
| NM_001270490.1 | NP_001257419.1 | |||
| NR_073022.1 | ||||
| AI921565.1 | ||||
| AK026558.1 | ||||
| AK091257.1 | 1 | 2344 | ||
| AK293386.1 | ||||
| BC000374.2 | AAH00374.1 | |||
| BC009708.2 | AAH09708.1 | |||
| BC021743.2 | AAH21743.1 | |||
| BM764512.1 | ||||
| CD246847.1 | ||||
| CD389415.1 | ||||
| DQ403032.1 | ||||
| L11566.1 | AAA16329.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| dgv3617n100 | Chr.19:48532114 - 48643732 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map