Assay Details
Target Gene Details
Entrez Gene ID: | 8178 |
Gene Name: | elongation factor for RNA polymerase II |
Gene Aliases: |
C19orf17, ELL1, MEN, PPP1R68 |
Location: |
Chr.19:18442663-18522129 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELL | NM_006532.3 | NP_006523.1 | ||
| XM_011528330.2 | XP_011526632.2 | |||
| XM_017027335.1 | XP_016882824.1 | |||
| XM_017027336.1 | XP_016882825.1 | |||
| XM_017027337.1 | XP_016882826.1 | |||
| AB208794.1 | BAD92031.1 | |||
| AK225481.1 | ||||
| AK293105.1 | ||||
| BC049195.1 | AAH49195.1 | |||
| U16282.1 | AAA57120.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952420 | Chr.19:18117791 - 18476990 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map