Assay Details
Target Gene Details
Entrez Gene ID: | 26330 |
Gene Name: | glyceraldehyde-3-phosphate dehydrogenase, spermatogenic |
Gene Aliases: |
GAPD2, GAPDH-2, GAPDS, HEL-S-278, HSD-35 |
Location: |
Chr.19:35533338-35545319 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAPDHS | NM_014364.4 | NP_055179.1 | ||
| XM_017026579.1 | XP_016882068.1 | |||
| AJ005371.1 | CAA06501.1 | |||
| AK225727.1 | ||||
| AK314980.1 | ||||
| AY306129.1 | AAQ75383.1 | |||
| BC036373.2 | AAH36373.1 | |||
| BP370184.1 | ||||
| GQ472225.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953285 | Chr.19:35520199 - 35562498 on Build GRCh38 | Deletion |
|
| nsv833811 | Chr.19:35359355 - 35538633 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map