Assay Details
Target Gene Details
Entrez Gene ID: | 100507567 |
Gene Name: | RAB11B antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.19:8374376-8390691 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB11B-AS1 | NR_038237.1 | 1 | 468 | |
| BG031227.1 | 1 | 468 | ||
| BM983524.1 | 3 | 567 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv28519 | Chr.19:8389955 - 8391295 on Build GRCh38 | Loss |
|
| esv3643614 | Chr.19:8333390 - 8391885 on Build GRCh38 | Gain |
|

Genomic Map