Assay Details
Target Gene Details
Entrez Gene ID: | 9266 |
Gene Name: | cytohesin 2 |
Gene Aliases: |
ARNO, CTS18, CTS18.1, PSCD2, PSCD2L, SEC7L, Sec7p-L, Sec7p-like |
Location: |
Chr.19:48469208-48482314 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYTH2 | NM_004228.6 | 12 | 2385 | NP_004219.3 |
| NM_017457.5 | 13 | 2388 | NP_059431.1 | |
| AF318380.1 | AAL55887.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| nsv828572 | Chr.19:48399035 - 48497609 on Build GRCh38 | Loss |
|
| nsv518578 | Chr.19:48475068 - 48510834 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map