Assay Details
Target Gene Details
Entrez Gene ID: | 170463 |
Gene Name: | single stranded DNA binding protein 4 |
Gene Aliases: |
- |
Location: |
Chr.19:18418719-18434562 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SSBP4 | NM_001009998.3 | NP_001009998.1 | ||
| NM_032627.4 | NP_116016.1 | |||
| XM_005259790.3 | XP_005259847.1 | |||
| XM_006722665.3 | XP_006722728.1 | |||
| XM_006722666.2 | XP_006722729.1 | |||
| XM_006722668.2 | XP_006722731.1 | |||
| XM_017026431.1 | XP_016881920.1 | |||
| XM_017026432.1 | XP_016881921.1 | |||
| XM_017026433.1 | XP_016881922.1 | |||
| XM_017026434.1 | XP_016881923.1 | |||
| XM_017026435.1 | XP_016881924.1 | |||
| XM_017026436.1 | XP_016881925.1 | |||
| XM_017026437.1 | XP_016881926.1 | |||
| AK098429.1 | ||||
| BC000274.1 | AAH00274.1 | |||
| BF207000.1 | ||||
| BM851755.1 | ||||
| BX400247.2 | ||||
| CX873722.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952420 | Chr.19:18117791 - 18476990 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map