Assay Details
Target Gene Details
Entrez Gene ID: | 478 |
Gene Name: | ATPase Na+/K+ transporting subunit alpha 3 |
Gene Aliases: |
AHC2, ATP1A1, CAPOS, DYT12, RDP |
Location: |
Chr.19:41966582-41994276 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP1A3 | NM_001256213.1 | NP_001243142.1 | ||
| NM_001256214.1 | NP_001243143.1 | |||
| NM_152296.4 | NP_689509.1 | |||
| AK223569.1 | BAD97289.1 | |||
| AK293784.1 | ||||
| AK295078.1 | ||||
| AK295833.1 | ||||
| AK296557.1 | ||||
| AK307921.1 | ||||
| AK316069.1 | ||||
| BC009282.2 | AAH09282.1 | |||
| BC009394.2 | AAH09394.1 | |||
| BC015566.2 | AAH15566.1 | |||
| BX478337.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss |
|
| nsv953580 | Chr.19:41888230 - 41997948 on Build GRCh38 | Deletion |
|
| dgv3550n100 | Chr.19:41916194 - 42001892 on Build GRCh38 | Gain |
|
| nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map