Assay Details
Target Gene Details
Entrez Gene ID: | 5300 |
Gene Name: | peptidylprolyl cis/trans isomerase, NIMA-interacting 1 |
Gene Aliases: |
DOD, UBL5 |
Location: |
Chr.19:9835207-9849689 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIN1 | NM_006221.3 | NP_006212.1 | ||
| NR_038422.2 | ||||
| NR_038830.1 | ||||
| XM_011528068.2 | XP_011526370.1 | |||
| AF086512.1 | ||||
| AK097558.1 | ||||
| AK291074.1 | ||||
| BC002899.1 | AAH02899.1 | |||
| BC031971.1 | 1 | 431 | AAH31971.1 | |
| BG912418.1 | ||||
| BI827520.1 | ||||
| BI827672.1 | ||||
| BT019331.1 | AAV38138.1 | |||
| CD675162.1 | ||||
| CR407654.1 | CAG28582.1 | |||
| U49070.1 | AAC50492.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520683 | Chr.19:9807553 - 9968417 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map