Assay Details
Target Gene Details
Entrez Gene ID: | 64344 |
Gene Name: | hypoxia inducible factor 3 alpha subunit |
Gene Aliases: |
HIF-3A, HIF3-alpha-1, IPAS, MOP7, PASD7, bHLHe17 |
Location: |
Chr.19:46297046-46343433 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3583441 | Chr.19:46312040 - 46314442 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map