Assay Details
Target Gene Details
Entrez Gene ID: | 5714 |
Gene Name: | proteasome 26S subunit, non-ATPase 8 |
Gene Aliases: |
HEL-S-91n, HIP6, HYPF, Nin1p, Rpn12, S14, p31 |
Location: |
Chr.19:38374550-38383824 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMD8 | NM_002812.4 | NP_002803.2 | ||
| AK301771.1 | ||||
| AK304486.1 | ||||
| AK308809.1 | ||||
| BC001164.1 | AAH01164.3 | |||
| BC065006.1 | ||||
| BI560993.1 | ||||
| BQ684248.1 | ||||
| BT019688.1 | AAV38494.1 | |||
| D38047.1 | BAA07237.1 | |||
| EU668354.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953296 | Chr.19:38324061 - 38403060 on Build GRCh38 | Deletion |
|
| nsv833822 | Chr.19:38270108 - 38454465 on Build GRCh38 | Loss |
|
| nsv833823 | Chr.19:38364910 - 38545160 on Build GRCh38 | Loss |
|
| nsv470140 | Chr.19:38349499 - 38628854 on Build GRCh38 | Gain |
|
| nsv1064275 | Chr.19:38369225 - 38422611 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map