Assay Details
Target Gene Details
Entrez Gene ID: | 9266 |
Gene Name: | cytohesin 2 |
Gene Aliases: |
ARNO, CTS18, CTS18.1, PSCD2, PSCD2L, SEC7L, Sec7p-L, Sec7p-like |
Location: |
Chr.19:48469208-48482314 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYTH2 | NM_004228.6 | NP_004219.3 | ||
| NM_017457.5 | NP_059431.1 | |||
| XM_006723472.2 | XP_006723535.1 | |||
| XM_006723473.2 | XP_006723536.1 | |||
| AB209573.1 | 8 | 2521 | BAD92810.1 | |
| AF318380.1 | 1 | 1431 | AAL55887.1 | |
| AK292405.1 | ||||
| BC004361.1 | AAH04361.1 | |||
| BC038713.1 | AAH38713.1 | |||
| U59752.1 | AAB17532.1 | |||
| U70728.1 | AAB09591.1 | |||
| X99753.1 | CAA68084.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| nsv828572 | Chr.19:48399035 - 48497609 on Build GRCh38 | Loss |
|
| nsv518578 | Chr.19:48475068 - 48510834 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map