Assay Details
Target Gene Details
Entrez Gene ID: | 26664 |
Gene Name: | olfactory receptor family 7 subfamily C member 1 |
Gene Aliases: |
CIT-HSP-146E8, HSTPCR86P, OR19-5, OR7C4, TPCR86 |
Location: |
Chr.19:14797550-14835376 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR7C1 | XM_005259853.3 | XP_005259910.1 | ||
| XM_011527915.2 | XP_011526217.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2663270 | Chr.19:14804467 - 14812142 on Build GRCh38 | Deletion |
|
| nsv963011 | Chr.19:14799221 - 14805194 on Build GRCh38 | Duplication |
|
| nsv442454 | Chr.19:14804829 - 14812071 on Build GRCh38 | Loss |
|
| esv3583258 | Chr.19:14804102 - 14815683 on Build GRCh38 | Loss |
|
| esv3643752 | Chr.19:14804503 - 14812143 on Build GRCh38 | Loss |
|
| esv3643751 | Chr.19:14797644 - 14810922 on Build GRCh38 | Loss |
|
| esv3893167 | Chr.19:14610447 - 14950404 on Build GRCh38 | Gain |
|
| nsv514890 | Chr.19:14805077 - 14811820 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map