Assay Details
Target Gene Details
Entrez Gene ID: | 645 |
Gene Name: | biliverdin reductase B |
Gene Aliases: |
BVRB, FLR, HEL-S-10, SDR43U1 |
Location: |
Chr.19:40447784-40465818 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BLVRB | NM_000713.2 | NP_000704.1 | ||
| AK312137.1 | ||||
| BC109371.1 | AAI09372.1 | |||
| BU528444.1 | ||||
| D26308.1 | BAA05370.1 | |||
| D32143.1 | BAA06874.1 | |||
| EU794593.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833831 | Chr.19:40290833 - 40454958 on Build GRCh38 | Loss |
|
| esv2658680 | Chr.19:40447040 - 40449286 on Build GRCh38 | Deletion |
|
| esv3644349 | Chr.19:40447641 - 40453028 on Build GRCh38 | Loss |
|
| esv3644348 | Chr.19:40447031 - 40449320 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map