Assay Details
Target Gene Details
Entrez Gene ID: | 7386 |
Gene Name: | ubiquinol-cytochrome c reductase, Rieske iron-sulfur polypeptide 1 |
Gene Aliases: |
RIP1, RIS1, RISP, UQCR5 |
Location: |
Chr.19:29207260-29213229 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UQCRFS1 | NM_006003.2 | NP_005994.2 | ||
| AK291134.1 | ||||
| BC000649.1 | AAH00649.1 | |||
| BC010035.2 | AAH10035.1 | |||
| BC067832.1 | AAH67832.1 | |||
| BF210184.1 | ||||
| BF979273.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1061604 | Chr.19:28373028 - 29773397 on Build GRCh38 | Loss |
|
| dgv1034e212 | Chr.19:29198360 - 29215107 on Build GRCh38 | Loss |
|
| nsv2452 | Chr.19:29198755 - 29234024 on Build GRCh38 | Insertion |
|
| nsv579240 | Chr.19:29185365 - 29401390 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map