Assay Details
Target Gene Details
Entrez Gene ID: | 10452 |
Gene Name: | translocase of outer mitochondrial membrane 40 |
Gene Aliases: |
C19orf1, D19S1177E, PER-EC1, PEREC1, TOM40 |
Location: |
Chr.19:44891220-44903689 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOMM40 | NM_001128916.1 | NP_001122388.1 | ||
| NM_001128917.1 | NP_001122389.1 | |||
| NM_006114.2 | NP_006105.1 | |||
| XM_005258411.3 | XP_005258468.1 | |||
| AF043250.1 | AAC82342.1 | |||
| AF316398.1 | AAL46624.1 | |||
| AF316399.1 | AAL46625.1 | |||
| AF316401.1 | AAL46626.1 | |||
| AF316402.1 | AAL46627.1 | |||
| BC001779.1 | AAH01779.1 | |||
| BC006413.1 | AAH06413.1 | |||
| BC012134.1 | AAH12134.1 | |||
| BC017224.2 | AAH17224.1 | |||
| BC047528.1 | AAH47528.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509747 | Chr.19:44877936 - 44943426 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs371928862] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map