Assay Details
Target Gene Details
Entrez Gene ID: | 1346 |
Gene Name: | cytochrome c oxidase subunit 7A1 |
Gene Aliases: |
COX7A, COX7AH, COX7AM |
Location: |
Chr.19:36150922-36152869 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COX7A1 | NM_001864.3 | NP_001855.1 | ||
| AK312091.1 | ||||
| BC002757.2 | AAH02757.1 | |||
| BT006924.1 | AAP35570.1 | |||
| CR542120.1 | CAG46917.1 | |||
| HE647857.1 | ||||
| M83186.1 | AAA52166.1 |
More Information
Set Membership: |
|

Genomic Map