Assay Details
Target Gene Details
Entrez Gene ID: | 390877 |
Gene Name: | adenylate kinase isoenzyme 1-like |
Gene Aliases: |
- |
Location: |
Chr.19:6393261-6412404 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC390877 | XM_017026805.1 | XP_016882294.1 |
More Information
Set Membership: |
|

Genomic Map