Assay Details
Target Gene Details
Entrez Gene ID: | 10683 |
Gene Name: | delta like canonical Notch ligand 3 |
Gene Aliases: |
SCDO1 |
Location: |
Chr.19:39498917-39508481 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLL3 | NM_016941.3 | NP_058637.1 | ||
| NM_203486.2 | NP_982353.1 | |||
| AK075302.1 | BAC11535.1 | |||
| AK308460.1 | ||||
| BC000218.2 | AAH00218.1 | |||
| DB035825.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523856 | Chr.19:39422918 - 39506832 on Build GRCh38 | Loss |
|
| nsv953573 | Chr.19:39444461 - 39522960 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map