Assay Details
Target Gene Details
Entrez Gene ID: | 390928 |
Gene Name: | acid phosphatase 7, tartrate resistant (putative) |
Gene Aliases: |
PAPL, PAPL1 |
Location: |
Chr.19:39083011-39111489 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACP7 | NM_001004318.2 | NP_001004318.2 | ||
| XM_011526966.2 | XP_011525268.1 | |||
| XM_011526967.2 | XP_011525269.1 | |||
| XM_011526969.1 | XP_011525271.1 | |||
| XM_017026807.1 | XP_016882296.1 | |||
| XM_017026808.1 | XP_016882297.1 | |||
| AK131245.1 | BAD18425.1 | |||
| BC136721.1 | ||||
| BC136722.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833829 | Chr.19:39079041 - 39240137 on Build GRCh38 | Loss |
|
| nsv579508 | Chr.19:39043114 - 39137410 on Build GRCh38 | Gain |
|
| dgv3528n100 | Chr.19:39076588 - 39134150 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map